Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198292.3 | NP_938034.2 | 3, 4 | 1941 | ||
GenBank mRNA | AK029537.1 | - | 3, 4 | 1948 | ||
AK129434.1 | - | 3, 4 | 1941 | |||
AK168397.1 | - | 3, 4 | 1951 | |||
BC057406.1 | - | 2, 3 | 351 | 5687258 |