Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfsd7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145447.2 | NP_663422.1 | 6 | 1570 | ||
GenBank mRNA | AK145696.1 | - | 6 | 1527 | ||
AY260574.1 | - | 6 | 1570 | |||
AY260575.1 | - | 6 | 1570 | |||
AY260576.1 | - | 6 | 1570 | |||
BC011209.1 | - | 6 | 1570 | 4190027 |