Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ism2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290302.1 | NP_001277231.1 | 3 | 517 | ||
GenBank mRNA | BU054611.1 | - | 2 | 322 |