Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pacs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081170.1 | NP_001074639.1 | 13, 14 | 1448 | ||
NM_001291445.1 | NP_001278374.1 | 12, 13 | 1358 | |||
GenBank mRNA | AK036581.1 | - | 12, 13 | 1334 | ||
AK122326.1 | - | 11, 12 | 1137 |