Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgfbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009370.2 | NP_033396.1 | 4 | 751 | ||
GenBank mRNA | AK015337.1 | - | 3 | 687 | ||
AK154381.1 | - | 3 | 514 | |||
BC063260.1 | - | 4 | 751 | 6409765 | ||
CT010393.1 | - | 4 | 744 | |||
D25540.1 | - | 4 | 737 | |||
D28526.1 | - | 4 | 744 |