Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgtp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011579.3 | NP_035709.3 | 2 | 1298 | ||
GenBank mRNA | AK089836.1 | - | 2 | 1300 | ||
AK163978.1 | - | 2 | 1299 | |||
AK166353.1 | - | 2 | 1311 | |||
AK172473.1 | - | 2 | 1298 | |||
BC085259.1 | - | 1269 | 5389279 | |||
U15636.1 | - | 2 | 1142 |
Gene Symbol: Tgtp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145164.1 | NP_001138636.1 | 2 | 1305 | ||
GenBank mRNA | AK088858.1 | - | 2 | 1299 | ||
AK172580.1 | - | 2 | 1298 | |||
L38444.1 | - | 2 | 1136 |