Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfyve16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173392.4 | NP_775568.1 | 18 | 4681 | ||
GenBank mRNA | AK080779.1 | - | 14 | 2491 | ||
AK122241.1 | - | 18 | 4662 | |||
AK139138.1 | - | 3 | 306 | |||
BC042669.1 | - | 18 | 4679 | 4235024 |