Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tiam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145886.1 | NP_001139358.1 | 16 | 3391 | ||
NM_009384.3 | NP_033410.2 | 16 | 3426 | |||
GenBank mRNA | AK147392.1 | - | 16 | 3392 | ||
AK161477.1 | - | 7 | 733 | |||
AK162945.1 | - | 3 | 380 | |||
U05245.1 | - | 16 | 3429 |