Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170669.1 | NP_001164140.1 | 11 | 1374 | ||
NM_172263.2 | NP_758467.2 | 12 | 1107 | |||
GenBank mRNA | AK036090.1 | - | 14 | 1444 | ||
AK043680.1 | - | 11 | 1367 | |||
AK045991.1 | - | 11 | 1376 | |||
AK083166.1 | - | 11 | 1012 | |||
BC032286.1 | - | 12 | 1119 | 5367055 | ||
EF644173.1 | - | 14 | 1119 |