Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172592.2 | NP_766180.1 | 4, 5 | 1183 | ||
GenBank mRNA | AK028794.1 | - | 1 | 2719 | ||
AK033354.1 | - | 4, 5 | 1183 | |||
AK037115.1 | - | 4, 5 | 1159 | |||
AK039311.1 | - | 6, 7 | 918 | |||
AK044086.1 | - | 3, 4 | 487 | |||
AK052265.1 | - | 6, 7 | 1052 | |||
AK054414.1 | - | 5 | 1822 | |||
BC063761.1 | - | 7, 8 | 928 | |||
BC070460.1 | - | 4, 5 | 556 | 30548562 |