Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044384.1 | NP_001037849.1 | 5 | 583 | ||
NM_001294280.2 | NP_001281209.1 | 4 | 503 | |||
NM_011593.2 | NP_035723.2 | 5 | 580 | |||
GenBank mRNA | AA184230.1 | - | 3 | 349 | ||
AY622853.1 | - | 2 | 117 | |||
BC008107.1 | - | 5 | 485 | 3158222 | ||
BC034260.1 | - | 5 | 485 | 3158222 | ||
BC051260.1 | - | 5 | 465 | 30056565 | ||
BU756519.1 | - | 2 | 319 | |||
M17243.1 | - | 5 | 446 | |||
X04684.1 | - | 5 | 522 |