Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011594.3 | NP_035724.2 | 2 | 525 | ||
GenBank mRNA | AK032818.1 | - | 2 | 439 | ||
AK081522.1 | - | 2 | 621 | |||
AK144236.1 | - | 2 | 440 | |||
AK194058.1 | - | 1 | 79 | |||
AY622854.1 | - | 1 | 55 | |||
BC049126.1 | - | 2 | 422 | 6821553 | ||
M82858.1 | - | 2 | 420 | |||
M93954.1 | - | 2 | 212 | |||
X62622.1 | - | 2 | 430 |