Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm10044 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK041589.1 | - | 2 | 284 | ||
AK158007.1 | - | 2 | 270 | |||
BB623994.1 | - | 2 | 247 |