Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081247.1 | NP_001074716.1 | 3 | 411 | ||
GenBank mRNA | AK039440.1 | - | 3 | 424 | ||
AK143315.1 | - | 3 | 361 | |||
AK170475.1 | - | 3 | 418 | |||
BC053071.1 | - | 3 | 374 | |||
BC151002.1 | - | 3 | 321 | 9087915 | ||
BC151168.1 | - | 3 | 319 | 9088057 | ||
BC151169.1 | - | 3 | 321 | 9088058 |