Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285529.1 | NP_001272458.1 | 9 | 1509 | ||
NM_001285530.1 | NP_001272459.1 | 7 | 1290 | |||
NM_011599.5 | NP_035729.3 | 9 | 1512 | |||
GenBank mRNA | AK048515.1 | - | 9 | 1499 | ||
AK052961.1 | - | 7 | 1613 | |||
AK082499.1 | - | 9 | 1478 | |||
AY155200.1 | - | 3 | 145 | |||
BC057573.1 | - | 9 | 1339 | 6401650 | ||
U61362.1 | - | 9 | 678 |