Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285529.1 | NP_001272458.1 | 18 | 2998 | ||
NM_001285530.1 | NP_001272459.1 | 16 | 2779 | |||
NM_011599.5 | NP_035729.3 | 18 | 3001 | |||
GenBank mRNA | AK048515.1 | - | 18 | 2985 | ||
AK052961.1 | - | 16 | 3102 | |||
AK082499.1 | - | 18 | 2967 | |||
AY155195.1 | - | 18 | 2160 | |||
AY155197.1 | - | 16 | 1937 | |||
AY155198.1 | - | 14 | 1800 | |||
AY155200.1 | - | 10 | 1088 | |||
BC057573.1 | - | 18 | 2828 | 6401650 | ||
U61362.1 | - | 18 | 2167 |