Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285529.1 | NP_001272458.1 | 2 | 929 | ||
NM_001285530.1 | NP_001272459.1 | 2 | 929 | |||
NM_001285531.1 | NP_001272460.1 | 2 | 929 | |||
NM_001285532.1 | NP_001272461.1 | 2 | 929 | |||
NM_011599.5 | NP_035729.3 | 2 | 929 | |||
GenBank mRNA | AK044642.1 | - | 2 | 931 | ||
AK046402.1 | - | 2 | 897 | |||
AK048515.1 | - | 2 | 915 | |||
AK076750.1 | - | 2 | 311 | |||
AK082499.1 | - | 2 | 895 | |||
AY155195.1 | - | 2 | 88 | |||
AY155196.1 | - | 2 | 898 | |||
AY155197.1 | - | 2 | 87 | |||
AY155198.1 | - | 2 | 88 | |||
AY155199.1 | - | 2 | 88 | |||
BC057573.1 | - | 2 | 759 | 6401650 | ||
U61362.1 | - | 2 | 95 |