Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tle4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF229633.1 | - | 2 | 74 | ||
AK017548.1 | - | 2 | 637 | |||
AK134989.1 | - | 2 | 705 | |||
BC086631.1 | - | 3 | 618 | |||
BY073734.1 | - | 2 | 146 | |||
CD549374.1 | - | 2 | 49 |