Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tlx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021901.3 | NP_068701.1 | 1 | 746 | ||
GenBank mRNA | BC018246.1 | - | 1 | 746 | 4164519 |