Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019634.2 | NP_062608.2 | 2 | 170 | ||
GenBank mRNA | AF052492.1 | - | 2 | 118 | ||
AK004776.1 | - | 2 | 130 | |||
AK147306.1 | - | 2 | 132 | |||
AK147404.1 | - | 2 | 134 | |||
AK195558.1 | - | 2 | 142 | |||
BC050153.1 | - | 2 | 118 | |||
BC058603.1 | - | 2 | 126 | 6529011 | ||
BY238972.1 | - | 2 | 170 | |||
D26483.1 | - | 2 | 118 |