Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033272.1 | NP_001028444.1 | 6, 7 | 2656 | ||
GenBank mRNA | AK151394.1 | - | 3, 4 | 536 | ||
AK220313.1 | - | 3, 4 | 378 | |||
BC035942.1 | - | 4, 5 | 674 | |||
BC138455.1 | - | 6, 7 | 2613 | 8861475 | ||
BC145285.1 | - | 6, 7 | 2613 | 9053839 | ||
CD352148.1 | - | 3, 4 | 420 |