Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmpo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080129.2 | NP_001073598.1 | 1 | 414 | ||
NM_001080130.2 | NP_001073599.1 | 1 | 414 | |||
NM_001080131.2 | NP_001073600.1 | 1 | 414 | |||
NM_001080132.2 | NP_001073601.1 | 1 | 414 | |||
NM_011605.3 | NP_035735.2 | 1 | 414 | |||
GenBank mRNA | AK141845.1 | - | 1 | 364 | ||
AK144642.1 | - | 1 | 374 | |||
AK146903.1 | - | 1 | 380 | |||
AK150305.1 | - | 1 | 362 | |||
AK150515.1 | - | 1 | 349 | |||
AK151936.1 | - | 1 | 366 | |||
AK152737.1 | - | 1 | 366 | |||
AK153281.1 | - | 1 | 349 | |||
AK156206.1 | - | 1 | 364 | |||
AK165282.1 | - | 1 | 364 | |||
AK165851.1 | - | 1 | 371 | |||
BC141062.1 | - | 1 | 132 | 9055674 | ||
CJ071529.1 | - | 1 | 415 | |||
U39073.1 | - | 1 | 317 | |||
U39074.1 | - | 1 | 345 | |||
U39075.1 | - | 1 | 345 | |||
U39076.1 | - | 1 | 345 | |||
U39077.1 | - | 1 | 345 | |||
U39078.1 | - | 1 | 328 |