Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289895.1 | NP_001276824.1 | 24 | 4070 | ||
NM_027884.3 | NP_082160.3 | 26 | 4131 | |||
GenBank mRNA | AK004758.1 | - | 3 | 658 | ||
AK156648.1 | - | 3 | 715 | |||
BC052740.1 | - | 5 | 896 | |||
BC055076.1 | - | 3 | 708 |