Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016785.2 | NP_058065.2 | 5, 6 | 564 | ||
GenBank mRNA | AF037043.1 | - | 5, 6 | 473 | ||
AF037044.1 | - | 4, 5 | 424 | |||
AF046887.1 | - | 5, 6 | 550 | |||
AK002335.1 | - | 5, 6 | 471 | |||
BC021598.1 | - | 4, 5 | 451 | 5098857 | ||
CF106558.1 | - | 5, 6 | 486 |