Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trex1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012236.1 | NP_001012236.1 | 2 | 545 | ||
NM_011637.6 | NP_035767.4 | 2 | 559 | |||
GenBank mRNA | AF151106.3 | - | 2 | 545 | ||
AF319574.1 | - | 1 | 1259 | |||
AK009899.1 | - | 2 | 546 | |||
AK087832.1 | - | 2 | 546 | |||
AK171916.1 | - | 2 | 534 | |||
BC011133.1 | - | 2 | 566 | 4206812 | ||
CA323364.1 | - | 2 | 699 | |||
CO041929.1 | - | 1 | 524 |