Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsks | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077591.1 | NP_001071059.1 | 8 | 1234 | ||
NM_011651.2 | NP_035781.2 | 8 | 1261 | |||
GenBank mRNA | AF025310.2 | - | 8 | 1241 | ||
BC118618.1 | - | 8 | 1213 | 40107271 | ||
BC121827.1 | - | 8 | 1213 | 40107270 |