Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sumo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009460.2 | NP_033486.1 | 4 | 362 | ||
GenBank mRNA | AF033353.1 | - | 4 | 337 | ||
AK002536.1 | - | 4 | 332 | |||
AK011074.1 | - | 4 | 331 | |||
AK089081.1 | - | 4 | 311 | |||
AK159366.1 | - | 4 | 333 | |||
AK220052.1 | - | 3 | 226 | |||
BC082566.1 | - | 4 | 337 | 30602751 | ||
BC083158.1 | - | 4 | 335 | 6478032 |