Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uncx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013702.3 | NP_038730.1 | 2 | 431 | ||
GenBank mRNA | AF247550.1 | - | 2 | 420 | ||
AJ001116.1 | - | 2 | 391 | |||
AK082472.1 | - | 2 | 432 | |||
BC051973.1 | - | 2 | 393 | 5718115 | ||
S83543.1 | - | 379 | ||||
Z96107.1 | - | 2 | 403 |