Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285517.1 | NP_001272446.1 | 7, 8 | 1184 | ||
NM_001285518.1 | NP_001272447.1 | 7, 8 | 1178 | |||
NM_001285519.1 | NP_001272448.1 | 7, 8 | 1175 | |||
NM_009473.3 | NP_033499.1 | 7, 8 | 1191 | |||
GenBank mRNA | AK077620.1 | - | 7, 8 | 1157 | ||
AK134489.1 | - | 7, 8 | 1141 | |||
AK156777.1 | - | 7, 8 | 1159 | |||
BC066025.1 | - | 7, 8 | 1142 | 6829527 | ||
BC106148.1 | - | 6, 7 | 911 | |||
EU869275.2 | - | 4, 5 | 1170 | |||
U09419.1 | - | 6, 7 | 1009 |