Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285517.1 | NP_001272446.1 | 9 | 1366 | ||
NM_001285518.1 | NP_001272447.1 | 9 | 1360 | |||
NM_001285519.1 | NP_001272448.1 | 9 | 1357 | |||
NM_009473.3 | NP_033499.1 | 9 | 1373 | |||
GenBank mRNA | AK077620.1 | - | 9 | 1339 | ||
AK134489.1 | - | 9 | 1323 | |||
AK156777.1 | - | 9 | 1341 | |||
AK177488.1 | - | 2 | 124 | |||
AK194055.1 | - | 2 | 124 | |||
BC066025.1 | - | 9 | 1324 | 6829527 | ||
BC106148.1 | - | 8 | 1093 | |||
EU869275.2 | - | 6 | 1352 | |||
U09419.1 | - | 8 | 1191 |