Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uty | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009484.2 | NP_033510.2 | 13, 14 | 1498 | ||
GenBank mRNA | AF057367.1 | - | 13, 14 | 1478 | ||
AK142344.1 | - | 15, 16 | 1582 | |||
AK142512.1 | - | 13, 14 | 1414 | |||
BC053061.1 | - | 13, 14 | 1481 |