Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uty | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009484.2 | NP_033510.2 | 15 | 2077 | ||
GenBank mRNA | AF057367.1 | - | 15 | 2057 | ||
AK142344.1 | - | 18 | 2215 | |||
AK142512.1 | - | 15 | 1993 | |||
AK155922.1 | - | 15 | 1822 | |||
AK162410.1 | - | 13 | 1650 | |||
AK164380.1 | - | 1 | 2014 | |||
AK164917.1 | - | 2 | 522 | |||
AK220463.1 | - | 16 | 2034 | |||
BC053061.1 | - | 15 | 2060 |