Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vamp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080557.1 | NP_001074026.1 | 2 | 238 | ||
NM_009496.3 | NP_033522.1 | 2 | 238 | |||
GenBank mRNA | AF007167.1 | - | 1 | 93 | ||
AK018783.1 | - | 2 | 238 | |||
AK037711.1 | - | 2 | 239 | |||
AK078156.1 | - | 2 | 239 | |||
BC049902.1 | - | 2 | 200 | 4500872 | ||
BC057587.1 | - | 2 | 217 | 6415847 | ||
U61751.1 | - | 2 | 184 |