Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011699.3 | NP_035829.1 | 4 | 316 | ||
GenBank mRNA | AF087695.1 | - | 4 | 316 | ||
AF173083.1 | - | 4 | 323 | |||
AK015399.1 | - | 4 | 335 | |||
AK078923.1 | - | 4 | 336 | |||
AK083299.1 | - | 4 | 340 | |||
AK128989.1 | - | 4 | 336 | |||
AK133531.1 | - | 4 | 325 | |||
AK135810.1 | - | 4 | 342 | |||
AK161076.1 | - | 4 | 297 | |||
AK162185.1 | - | 2 | 161 | |||
BC046966.1 | - | 4 | 316 | 6493071 | ||
BC052471.1 | - | 4 | 326 |