Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172606.2 | NP_766194.2 | 23, 24 | 2594 | ||
GenBank mRNA | AK028353.1 | - | 9, 10 | 1127 | ||
AK042980.1 | - | 8, 9 | 953 | |||
AK129169.1 | - | 11, 12 | 1337 | |||
AK132461.1 | - | 23, 24 | 2568 | |||
AK218077.1 | - | 2, 3 | 223 | |||
BC037454.1 | - | 9, 10 | 1148 | |||
BC048816.1 | - | 10, 11 | 1234 | |||
BC059190.1 | - | 9, 10 | 1122 |