Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naprt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172607.3 | NP_766195.2 | 5, 6 | 927 | ||
GenBank mRNA | AK033661.1 | - | 5, 6 | 930 | ||
AK049157.1 | - | 5, 6 | 676 | |||
AK159127.1 | - | 5, 6 | 726 | |||
AK170518.1 | - | 5, 6 | 726 | |||
AY214331.1 | - | 5, 6 | 930 | |||
BC058800.1 | - | 5, 6 | 697 | 6476979 |