Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eppk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144848.2 | NP_659097.2 | 7973 | |||
GenBank mRNA | AK041683.1 | - | 1 | 1254 | ||
AY312170.1 | - | 7973 | ||||
BC026387.1 | - | 1 | 249 |