Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5031439G07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033273.2 | NP_001028445.1 | 7 | 994 | ||
GenBank mRNA | AK138575.1 | - | 6 | 979 | ||
AK149782.1 | - | 7 | 995 | |||
AK170532.1 | - | 7 | 1027 | |||
AK173065.1 | - | 7 | 752 |