Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016852.2 | NP_058548.1 | 1 | 77 | ||
GenBank mRNA | AK004824.1 | - | 1 | 79 | ||
AK043037.1 | - | 1 | 51 | |||
AK079300.1 | - | 1 | 67 | |||
AK146572.1 | - | 1 | 79 | |||
AK150018.1 | - | 1 | 52 | |||
AK151368.1 | - | 1 | 69 | |||
AK162810.1 | - | 1 | 52 | |||
AK170317.1 | - | 1 | 78 | |||
AK170474.1 | - | 1 | 80 | |||
BC031908.1 | - | 1 | 40 | 5365118 | ||
BC055058.1 | - | 1 | 97 | 6529458 | ||
U40826.1 | - | 1 | 77 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK044015.1 | - | 67 | Chr. - - [-] on Build |