Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pphln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083114.1 | NP_001076583.1 | 6 | 651 | ||
NM_001285863.1 | NP_001272792.1 | 6 | 780 | |||
NM_001285864.1 | NP_001272793.1 | 5 | 559 | |||
NM_146062.3 | NP_666174.2 | 8 | 915 | |||
NM_175363.4 | NP_780572.1 | 7 | 708 | |||
GenBank mRNA | AK081606.1 | - | 7 | 701 | ||
AK150904.1 | - | 7 | 659 | |||
AK158488.1 | - | 6 | 781 | |||
AK160138.1 | - | 7 | 681 | |||
AK168443.1 | - | 6 | 653 | |||
AK206710.1 | - | 3 | 250 | |||
AK211919.1 | - | 3 | 250 | |||
BC031486.1 | - | 8 | 868 | 3157250 | ||
BC150854.2 | - | 5 | 555 | 9087765 | ||
CF734096.1 | - | 5 | 539 |