Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif4h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033561.1 | NP_291039.1 | 7 | 656 | ||
GenBank mRNA | AK045004.1 | - | 7 | 658 | ||
AK088818.1 | - | 7 | 668 | |||
AK122194.1 | - | 5 | 3878 | |||
AK132034.1 | - | 7 | 676 | |||
AK135390.1 | - | 7 | 657 | |||
AK136999.1 | - | 6 | 583 | |||
AK149841.1 | - | 7 | 642 | |||
AK150088.1 | - | 7 | 665 | |||
AK166567.1 | - | 7 | 657 | |||
AK168872.1 | - | 6 | 615 | |||
AK169312.1 | - | 7 | 639 | |||
AK169353.1 | - | 7 | 657 | |||
AK169407.1 | - | 6 | 615 | |||
AK183007.1 | - | 122 | ||||
AK188319.1 | - | 3 | 252 | |||
AK188850.1 | - | 2 | 108 | |||
AK210675.1 | - | 2 | 108 | |||
AK219562.1 | - | 2 | 108 | |||
BC014796.1 | - | 6 | 610 | 3962104 |