Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgef3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177810.1 | NP_001171281.1 | 25 | 2773 | ||
NM_001177811.1 | NP_001171282.1 | 24 | 2722 | |||
NM_144850.2 | NP_659099.2 | 25 | 2749 | |||
GenBank mRNA | AK034265.1 | - | 12 | 1113 | ||
AK044566.1 | - | 24 | 3521 | |||
AK142594.1 | - | 25 | 2667 | |||
AK162003.1 | - | 7 | 769 | |||
AK169430.1 | - | 24 | 2721 | |||
BC020311.2 | - | 7 | 591 | |||
BC020532.1 | - | 25 | 2761 | 4236136 |