Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Soat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146064.1 | NP_666176.1 | 14 | 1531 | ||
GenBank mRNA | AF078751.1 | - | 14 | 1501 | ||
BC025931.1 | - | 14 | 1531 | 5099063 |