Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpped1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146067.3 | NP_666179.2 | 3 | 690 | ||
GenBank mRNA | AK029379.1 | - | 3 | 614 | ||
AK032288.1 | - | 3 | 653 | |||
AK041329.1 | - | 3 | 663 | |||
AK049721.1 | - | 3 | 691 | |||
AK080876.1 | - | 3 | 648 | |||
AK133977.1 | - | 3 | 684 | |||
BC025476.1 | - | 3 | 611 | |||
BC034347.1 | - | 3 | 332 | 2654874 |