Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spidr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146068.4 | NP_666180.2 | 10, 11 | 1679 | ||
GenBank mRNA | AK044836.1 | - | 10, 11 | 1662 | ||
AK052832.1 | - | 10, 11 | 1642 | |||
AK075871.1 | - | 10, 11 | 1644 | |||
AK146053.1 | - | 10, 11 | 1679 | |||
AK166448.1 | - | 5, 6 | 974 | |||
AK170784.1 | - | 6, 7 | 1311 | |||
AK172902.1 | - | 10, 11 | 1632 | |||
BC026877.1 | - | 12 | ||||
BC046558.1 | - | 4, 5 | 683 | |||
BC069859.1 | - | 1, 2 | 232 | |||
BC085258.1 | - | 1, 2 | 50 | |||
BC089026.1 | - | 10, 11 | 1616 | 30615298 |