Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spidr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146068.4 | NP_666180.2 | 13 | 2017 | ||
GenBank mRNA | AK044836.1 | - | 13 | 2000 | ||
AK052832.1 | - | 13 | 1980 | |||
AK075871.1 | - | 13 | 1982 | |||
AK146053.1 | - | 13 | 2017 | |||
AK166448.1 | - | 8 | 1312 | |||
AK170784.1 | - | 9 | 1649 | |||
AK172902.1 | - | 13 | 1970 | |||
BC026877.1 | - | 3 | 351 | |||
BC046558.1 | - | 7 | 1021 | |||
BC069859.1 | - | 4 | 570 | |||
BC085258.1 | - | 4 | 388 | |||
BC089026.1 | - | 13 | 1954 | 30615298 |