Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013842.3 | NP_038870.2 | 4 | 833 | ||
GenBank mRNA | AF027963.3 | - | 4 | 507 | ||
AK007724.1 | - | 4 | 494 | |||
AK145241.1 | - | 4 | 493 | |||
AK204892.1 | - | 1 | 29 | |||
AK207925.1 | - | 1 | 18 | |||
AK211765.1 | - | 1 | 18 | |||
AK213043.1 | - | 1 | 18 | |||
BC008153.1 | - | 4 | 448 | |||
BC029197.1 | - | 4 | 488 | 2648085 |