Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2410015M20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153152.3 | NP_694792.1 | 4 | 380 | ||
GenBank mRNA | AK075942.1 | - | 4 | 311 | ||
AK075961.1 | - | 4 | 381 | |||
AY091637.1 | - | 4 | 323 | |||
BC049674.1 | - | 201 | ||||
BC056164.1 | - | 4 | 286 | 5713062 |