Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177638.4 | NP_808306.1 | 3 | 287 | ||
GenBank mRNA | AK077528.1 | - | 3 | 219 | ||
AK167279.1 | - | 3 | 217 | |||
BC024462.1 | - | 3 | 287 | 4975562 |