Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081034.1 | NP_001074503.1 | 18 | 2237 | ||
GenBank mRNA | AK132635.1 | - | 2 | 213 | ||
BC027802.1 | - | 7 | 814 | |||
BC049946.1 | - | 2 | 165 | |||
BC055343.1 | - | 9 | 979 | |||
BC110686.1 | - | 18 | 1976 | |||
BC117884.1 | - | 18 | 1983 | 40096054 | ||
BC117885.1 | - | 18 | 1987 | 40096055 | ||
BC128479.1 | - | 18 | 1987 | 40096053 |